Penicillin G, Reference Standards
Leverandør: USP
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1502009 1502508 1502552 1502701
1502009.EA 2790 DKK
1502009. 1502508. 1502552. 1502701.
Penicillin G, Reference Standards
Standarder Økologiske standarder
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certificeringer: US Pharmacopea Reference Standard
Bestil nu
Specifikationer
Beskrivelse Penicillin G Benzathine Penicillin G Potassium Penicillin G Procaine Penicillin G Sodium
Environmentally Preferable
CAS-nr 41372-02-5 113-98-4 6130-64-9 69-57-8
Formel C₄₈H₆₄N₆O₁₂S₂ C₂₉H₄₀N₄O₇S