Safety winter boots, lace-up, SnowCruiser
Leverandør: MILLE S.A.S
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
CRRS5/48 CRRS5/40 CRRS5/39 CRRS5/41 CRRS5/42 CRRS5/43 CRRS5/44 CRRS5/45 CRRS5/46 CRRS5/47 CRRS5/38
MILECRRS5/48EA 2430 DKK
MILECRRS5/48 MILECRRS5/40 MILECRRS5/39 MILECRRS5/41 MILECRRS5/42 MILECRRS5/43 MILECRRS5/44 MILECRRS5/45 MILECRRS5/46 MILECRRS5/47 MILECRRS5/38
Safety winter boots, lace-up, SnowCruiser
Sko og støvler
Safety winter boots made from a combination of PU/Cryoflex® upper and high quality, water-repellent leather leg plus a cold resistant, Cryoflex® outsole (slip resistant due to deep inverted studs).
  • Thermal resistance and good vapour permeability against cold temperatures up to –50 °C
  • Genuine lamb’s wool lining
  • Walkontan® shock-absorbent heel
  • Lacing system with waterproof tongue
  • Removable Dry-Tex™ boot sock, made from five different materials, ensures a very high level of insulation
  • Boot sock features a VELCRO® fastener, can be used as a normal boot sock; washable at 40 °C
  • Composite XXL toecap improves inside comfort, even with the Dry-Tex™ boot sock
  • Textile, puncture resistant midsole
Suitable for cold and wet environments.
Colour: Grey/blue
Certificeringer: EN ISO 20345; S5, SRC.