ESD safety clogs for ladies, Azalée, Flowergrip®
Leverandør: MILLE S.A.S
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
AZAT1/36 AZAT1/37 AZAT1/38 AZAT1/39 AZAT1/40 AZAT1/41 AZNT1/40 AZNT1/35 AZNT1/36 AZNT1/37 AZAT1/35 AZNT1/41 AZNT1/39 AZNT1/38
MILEAZAT1/36EA 733 DKK
MILEAZAT1/36 MILEAZAT1/37 MILEAZAT1/38 MILEAZAT1/39 MILEAZAT1/40 MILEAZAT1/41 MILEAZNT1/40 MILEAZNT1/35 MILEAZNT1/36 MILEAZNT1/37 MILEAZAT1/35 MILEAZNT1/41 MILEAZNT1/39 MILEAZNT1/38
ESD safety clogs for ladies, Azalée, Flowergrip®
Sko og støvler
These light and comfortable anti-static safety clogs are made from antibacterial microfibre, Lycra® inserts and a PU sole.
  • Abrasion-resistant, antibacterial poromax® lining with high water-absorbing capacity for additional breathability
  • Composite toe protection cap (200 J)
  • Shock absorber heel
  • Washable with warm soapy water
Suitable for food processing industries, services, catering, administration, slippery and wet soils.
Colour: Black/white
Certificeringer: EN ISO 20345; IEC EN 61340-5-1 (ESD); SB, E, A, SRC