Quinidine, Reference Standards
Leverandør: USP
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1595000 1595509
1595000.EA 2930 DKK
1595000. 1595509.
Quinidine, Reference Standards
Standarder Økologiske standarder
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certificeringer: US Pharmacopea Reference Standard
Bestil nu
Specifikationer
Beskrivelse Quinidine Gluconate Quinidine Sulfate
Environmentally Preferable
CAS-nr 7054-25-3 6591-63-5
Formel C₂₆H₃₆N₂O₉ C₄₀H₅₄N₄O₁₀S
UN No. 2811