Chemical resistant overalls, 4532+
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
4532PLUSBM 4532PLUSBXL 4532PLUSWS 4532PLUSWM 4532PLUSW3X 4532PLUSWL 4532PLUSWXL 4532PLUSBL 4532PLUSB3X 4532PLUSB2X 4532PLUSW2X
MMMA4532PLUSBMEA 2220 DKK
MMMA4532PLUSBM MMMA4532PLUSBXL MMMA4532PLUSWS MMMA4532PLUSWM MMMA4532PLUSW3X MMMA4532PLUSWL MMMA4532PLUSWXL MMMA4532PLUSBL MMMA4532PLUSB3X MMMA4532PLUSB2X MMMA4532PLUSW2X
Chemical resistant overalls, 4532+
Engangsbeklædning
These low-linting overalls made from soft and highly breathable SSMMS material provide enhanced liquid and dust protection. The 5-layer SSMMS material is specially treated to increase the repellence to certain oils and solvents enabling this product to be used in many applications not usually suitable for SMS-based overalls.
  • Excellent barrier against dry particles and certain limited liquid chemical splash (CE Type 5/6)
  • Alcohol and oil repellence treatment
  • Robust yet soft highly breathable SSMMS fabric has an anti-static treatment
  • Elasticated waist and ankles for convenience and freedom of movement and knitted cuffs for extra comfort
  • Two-way zipper with sealable storm flap for added on-off convenience and extra protection
  • Enhanced and reinforced triangular gusset panel (in blue)
  • Three-panel hood for optimal fit
Typical applications may include metal grinding or polishing, powder handling, oil related or greasy environments, cement manufacturing, painting and coating.
Certificeringer: Type 5/6, EN 1149, EN 1073-2; Cat. III