Turbidity standards, formazin
Leverandør: LGC Standards PROMOCHEM
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
RTCTURB1000100 RTCTURB05-500 RTCTURB100-500 RTCTURB1-500ML
PROORTCTURB1000100EA 1780 DKK
PROORTCTURB1000100 PROORTCTURB05-500 PROORTCTURB100-500 PROORTCTURB1-500ML
Turbidity standards, formazin
Standarder Turbiditets standarder
Turbidity calibration standard formazin solutions, RTC ranging from 0,5 to 4000 NTU.