Acrylamide/Bis-Acrylamide
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
J61505.AP J61505.K2
J61505.APEA 1760 DKK
J61505.AP J61505.K2
Acrylamide/Bis-Acrylamide
Elektroforese geler Agarose og Akrylamid
Premix of acrylamide with the common cross-linking reagent of final ratio 29:1.
  • Soluble in water
  • DNA sequencing gels and protein separation