Overhead stirrer tools, radial stirrers (IKA)
Leverandør: IKA
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
20001206 20001207 20001208
IKAA20001206EA 2840 DKK
IKAA20001206 IKAA20001207 IKAA20001208
Overhead stirrer tools, radial stirrers (IKA)
Omrørere Omrøreudstyr
These stirring blades can be used for aerating low viscosity media and homogenising solids in liquids.
  • Ideal for drawing the material to be mixed from the top and bottom
  • Generates a radial flow
  • Can be used at medium to high speeds