Chlorine test tablets, DPD TESTABS®
Leverandør: LaMotte Chemicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
6976AJ 6197A-J
LAMO6976AJEA 526 DKK
LAMO6976AJ LAMO6197A-J
Chlorine test tablets, DPD TESTABS®
Miljø test kit
These DPD tablet are used for determining free and combined chlorine in pool water.
  • DPD 1 quickly reveals the precise level of free chlorine
  • DPD 3 tablet measures the level of combined chlorine