Rods, ceramic, round, solid, AL23
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
32921.DM 32931.DR 32931.DT 32931.DW 32931.EH 32933.DR
32921.DMEA 248 DKK
32921.DM 32931.DR 32931.DT 32931.DW 32931.EH 32933.DR
Rods, ceramic, round, solid, AL23
Stænger
Pure Al₂O₃ Aluminium oxide, AL23.
Dense, excellent thermal and electrical resistance properties, corrosion resistant, permeable to microwaves.