VWR® Primera Signature, Printer Slides
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
631-1183EA 1450 DKK
631-1183 631-1184 631-1185
VWR® Primera Signature, Printer Slides
Objektglas Objektglas
Microscopy slides are designed with colour-frosted ends for haematoxylin and eosin (H&E) stain, immunohistochemistry (IHC) and hybridisation on frozen sections in histopathology examination.
  • Shelf life of 60 months
  • Pre-cleaned, without mark, print, or contamination risks
  • Marking with conventional labelling system, pencil, marker pens, and inkjet printers
Specially produced for all Primera printers.
Advarsel: Fragile items should be stored in dry and proper laboratory temperature conditions.