Padlocks, brass
Leverandør: Brady
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
051354 051355 051356 051357 051358 051359
BRDY051354EA 1260 DKK
BRDY051354 BRDY051355 BRDY051356 BRDY051357 BRDY051358 118-0517
Padlocks, brass
Sikkerhedslåse
These durable padlocks feature solid brass bodies and shackles that resist corrosion and sparks.
  • 6-pin cylinder offers more unique key cuts for better tamper resistance
  • Locks are protected by coloured vinyl sheaths making it easy to colour-code lockout procedures
  • Engraving on bottom and key
  • Padlocks which are keyed differently are available
Shackle clearance: 20,5 mm
Shackle diameter: 6,5 mm
Shackle height: 21,00 mm
Shackle width: 33,5 mm
Bestillingsoplysninger: Lock labels sold separately. Two keys are supplied with each padlock.