Butterfly valve lockouts
Leverandør: Brady
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
121504 121505
118-0521EA 249 DKK
118-0521 118-0522
Butterfly valve lockouts
Sikkerhedslåse
This lockout device can be used to lock a butterfly valve in the off position by clamping down on the handle so that it may not be engaged or moved.
  • Available in two sizes (small and large)
  • Maximum number of padlocks: 4
Small lockout: Accommodates valve handle thickness 3,20 to 63,50 mm
Large lockout: Accommodates valve handle thicknes 50,80 to 102 mm