Push button lockout devices
Leverandør: Brady
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
130819 130820 130821 134018 130822 130823 130824 139796 139793 139794 139795
BRDY130819EA 235 DKK
BRDY130819 118-0525 118-0526 118-0527 BRDY130822 BRDY130823 BRDY130824 BRDY139796 BRDY139793 BRDY139794 BRDY139795
Push button lockout devices
Sikkerhedslåse
These lockout devices prevent the unauthorised use of push button emergency devices.
  • With standard cover
  • Made from clear PC, with translucent red cover
  • Suitable for 16, 22 or 30 mm push buttons