ESD safety ankle boots, lace-up, uvex 2, vibram®, 6523
Leverandør: UVEX
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
6523.2/48 6523.2/49 6523.2/41 6523.2/42 6523.2/43 6523.2/44 6523.2/45 6523.2/40 6523.2/50 6523.2/51 6523.2/52 6523.2/36 6523.2/37 6523.2/38 6523.2/39 6523.2/35 6523.2/46 6523.2/47 6523.3/35 6523.3/36 6523.3/37 6523.3/38 6523.3/39 6523.3/40 6523.3/41 6523.3/42 6523.3/43 6523.3/44 6523.3/45 6523.3/46 6523.3/47 6523.3/48 6523.3/49 6523.3/50 6523.3/51 6523.3/52 6523.4/35 6523.4/36 6523.4/37 6523.4/38 6523.4/39 6523.4/40 6523.4/41 6523.4/42 6523.4/43 6523.4/44 6523.4/45 6523.4/46 6523.4/47 6523.4/48 6523.4/49 6523.4/50 6523.4/51 6523.4/52 6523.1/46 6523.1/47 6523.1/48 6523.1/49 6523.1/50 6523.1/51 6523.1/52 6523.1/35 6523.1/36 6523.1/37 6523.1/38 6523.1/39 6523.1/40 6523.1/41 6523.1/42 6523.1/43 6523.1/44 6523.1/45
114-8090EA 960 DKK
114-8090 114-8091 114-8083 114-8084 114-8085 114-8086 114-8087 114-8082 114-8092 114-8093 114-8094 114-8078 114-8079 114-8080 114-8081 114-8077 114-8088 114-8089 UVEX6523.3/35 UVEX6523.3/36 UVEX6523.3/37 UVEX6523.3/38 UVEX6523.3/39 UVEX6523.3/40 UVEX6523.3/41 UVEX6523.3/42 UVEX6523.3/43 UVEX6523.3/44 UVEX6523.3/45 UVEX6523.3/46 UVEX6523.3/47 UVEX6523.3/48 UVEX6523.3/49 UVEX6523.3/50 UVEX6523.3/51 UVEX6523.3/52 UVEX6523.4/35 UVEX6523.4/36 UVEX6523.4/37 UVEX6523.4/38 UVEX6523.4/39 UVEX6523.4/40 UVEX6523.4/41 UVEX6523.4/42 UVEX6523.4/43 UVEX6523.4/44 UVEX6523.4/45 UVEX6523.4/46 UVEX6523.4/47 UVEX6523.4/48 UVEX6523.4/49 UVEX6523.4/50 UVEX6523.4/51 UVEX6523.4/52 UVEX6523.1/46 UVEX6523.1/47 UVEX6523.1/48 UVEX6523.1/49 UVEX6523.1/50 UVEX6523.1/51 UVEX6523.1/52 UVEX6523.1/35 UVEX6523.1/36 UVEX6523.1/37 UVEX6523.1/38 UVEX6523.1/39 UVEX6523.1/40 UVEX6523.1/41 UVEX6523.1/42 UVEX6523.1/43 UVEX6523.1/44 UVEX6523.1/45
ESD safety ankle boots, lace-up, uvex 2, vibram®, 6523
Sko og støvler
Metal-free, lightweight, flexible, robust safety shoe made from breathable, water repellent leather, soft padded tongue and collar, plus a dual-density, slip-resistant Vibram® rubber sole, resistant to chemicals, oils and gasoline.
  • Climazone technology for improved comfort
  • Breathable distance mesh lining
  • Removable anti-static comfort insole with moisture control system; orthopaedic insoles possible
  • Virtually seam-free construction eliminates pressure points
  • Lightweight two-layer sole with excellent shock absorption and outstanding slip resistance
  • 100% metal-free xenova® toecap, non magnetic (200 J)
  • Injection-moulded PU scuff cap for additional protection
  • Flexible, metal-free, puncture resistant midsole
  • Fulfils ESD requirements: Resistance to ground inferior to 35 megaohms
For medium industrial applications including chemical, transport, logistics, energy and mechanical engineering industries.
Colour: Black/orange
Certificeringer: EN 61340 (ESD); DGUV112-191; EN ISO 20345:2011; S3, HI, HRO, SRC