Support belt, PW80
Leverandør: Portwest
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
PW80BKRL PW80BKRM PW80BKRS PW80BKRXL
118-0511EA 162 DKK
118-0511 118-0512 118-0513 118-0514
Support belt, PW80
Ergonomiske støtter Rygstøtter
This high performance belt made from 62% polyester and 38% latex offers full support to the wearer. It reinforces the spine, supports the abdomen and the lower back.
  • Adjustable for added comfort
  • Suitable for wearing under or over clothing
  • Four heavy-duty bonding strips with reinforced elastic for improved fit
Width: 20 cm.