Crucibles, Tammann, aluminum oxide
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
33052.KF 33053.KF 33055.KF 33056.KF 33058.KF
33052.KFEA 567 DKK
33052.KF 33053.KF 33055.KF 33056.KF 33058.KF
Crucibles, Tammann, aluminum oxide
Digler
These crucibles according to Tammann are manufactured from aluminium oxide. They display superior characteristics both at elevated temperature and at high frequencies, and are resistant to corrosive liquids and gases. Oxide ceramics are also inert to oxidation and are not subject to radiation damage.