Crucibles, cylindrical, flat base, aluminum oxide (Al-23)
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
32950.KF 32951.KF 32952.KF 32953.KF 32954.KF 32955.KF 32957.KF
32950.KFEA 690 DKK
32950.KF 32951.KF 32952.KF 32953.KF 32954.KF 32955.KF 32957.KF
Crucibles, cylindrical, flat base, aluminum oxide (Al-23)
Digler
These crucibles are manufactured from aluminium oxide. They display superior characteristics both at elevated temperature and at high frequencies, and are resistant to corrosive liquids and gases. Oxide ceramics are also inert to oxidation and are not subject to radiation damage.
  • 1950 °C (max service temp in air)