VWR®, Glass Stoppers for Standard Ground Joints
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
217-0887EA 92.3 DKK
217-0887 217-0888 217-0889 217-0890 217-0891 217-0892 217-0893 217-0894
VWR®, Glass Stoppers for Standard Ground Joints
Propper Glas
Borosilicate 3.3 glass.
  • Hollow, with hexagonal grip
  • Trip-tip to ensure every last drop of liquid returns into the vessel
  • Resistant to chemicals and thermal shock
Certificeringer: DIN 12252