General purpose gloves, Abratex Light Grip
Leverandør: HONEYWELL SAFETY
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
RGT888/7 RGT888/10 RGT888/8 RGT888/9
DALLRGT888/7EA 0 DKK
DALLRGT888/7 DALLRGT888/10 DALLRGT888/8 DALLRGT888/9
General purpose gloves, Abratex Light Grip
Handsker
These knitted gloves are made from 67% polyamide and 33% cotton and feature a dot coating on their palm.
  • Knitting technique helps to obtain a glove that conforms snugly to the hand, with good dexterity
  • Cotton interior slows down perspiration and increases comfort
  • The blend of two fibres, polyamide and cotton, ensures better abrasion and cutting resistance
  • Its white colour enhances the glove's hygiene and cleanliness properties
  • Knitted wrist (5 cm) with size related coloured edge
Ideal for fine handling and assembling operations.
Certificeringer: ENMore Product Information