VWR® B'Wipes 4000, Cleanroom wipes
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
115-1827EA 412 DKK
115-1827 115-1828
VWR® B'Wipes 4000, Cleanroom wipes
Papirservietter Servietter til Renrum Tør
Polyester, sealed-edge wipe for use in ISO 3 cleanrooms.
  • Single-knit, polyester wipe, with sealed edges (150 g/m²)
  • Laundered, with sealed edges, for superior cleanliness
  • Good mechanical resistance and absorption capacity
  • Ideal for cleaning and disinfecting surfaces in critical environments