VWR® B'Wipes 3000, Cleanroom wipes
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
115-1826EA 246 DKK
115-1826 115-1806 115-1805
VWR® B'Wipes 3000, Cleanroom wipes
Papirservietter Servietter til Renrum Tør
Polyester wipe for use in ISO 4 cleanrooms.
  • Single-knit, polyester wipe, with knife-cut edges (130 g/m²)
  • Laundered for better cleanliness
  • Good wiping efficiency and abrasion resistance
  • Ideal for general cleaning of all types of surfaces
  • Available in an economical version for floors