VWR® B'Wipes 1000, Cleanroom wipes
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
115-1804EA 165 DKK
115-1804 115-1803 115-1825
VWR® B'Wipes 1000, Cleanroom wipes
Papirservietter Servietter til Renrum Tør
Absorbent, non woven wipe for use in ISO 6 cleanrooms.
  • Non woven wipe, made of hydroentangled 55% cellulose and 45% polyester (68 g/m²)
  • High absorbency, strong and durable
  • Works well with IPA and other solvents
  • Ideal for spill control and general purpose wiping