Specimen collection and transport kits, Capitol Vial™
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
90111 92023
216-2138EA 1320 DKK
216-2138 216-2139
Specimen collection and transport kits, Capitol Vial™
Narkotikatest
Capitol Vial™ are specimen containers manufactured airtight, leak resistant and aseptic by process.
  • Twin valve design which creates a triple seal at the flip cap and vial interface
  • Superior leak resistant performance
  • Specimen samples are contained during transport and storage
Capitol Vial™ kits are sealed inside the collection cup with a foil top lid for transport (in accordance with IATA requirements) or other applications where a second sample is required.
Leveringsinformation: Capitol Vial™ kits contain 1×240 ml beaker with temperature indicatorMore Product Information