Post-mortem scalpel blades
Leverandør: SWANN MORTON
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2551 2561 2560 2552
233-0110EA 494 DKK
233-0110 233-0113 233-0112 233-0111
Post-mortem scalpel blades
Skalpeller Skalpelblade
A versatile and comprehensive range for morticians and pathologists for use in autopsy, histopathology and general post-mortem procedures.
  • PM40 range with traditional screw fitting
  • PM60 range with standard blade fitment
The PM40B and PM60B blades are 'bull nose' blades for added safety and protection against accidental sharps injuries.
Emballering: All blades are individually wrapped. The PM8 and PM40 stainless steel handles are supplied individually in a protective plastic wallet.