Test kits, photometric, Formaldehyde, LCK cuvette tests
Leverandør: Hach
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
LCK325 LCK425
705-3047EA 1020 DKK
705-3047 HACHLCK425
Test kits, photometric, Formaldehyde, LCK cuvette tests
Miljø test kit
Pre-dosed reagents for maximum safety.
  • Precise and reliable measurement values
  • Maximum safety for users, thanks to the closed system and low amounts of reagents
  • Convenient and error-free dosing of the reagents
  • Barcode label for automatic recognition in the photometer
  • Differentiation between tests and measuring ranges by means of colour-coding
Storage conditions: 15 to 25 °C
Certificeringer: LCK 425 according to standard ISO12460.