Gas purifiers
Leverandør: Trajan Scientific and Medical
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1035210 1035220 1035230 1035250 1035158 1035154 1035168 1035164
SGEA1035210EA 1410 DKK
SGEA1035210 SGEA1035220 SGEA1035230 SGEA1035250 SGEA1035158 SGEA1035154 SGEA1035168 SGEA1035164
Gas purifiers
Filtre Gasfiltre
Gas purifiers are designed to provide fast stabilisation times to reduce gas consumption and provide clean gas to GC and GC/MS systems.
  • Removal of oxygen, moisture and hydrocarbons that can impact analytical results
  • Fast stabilisation times
  • Improved trace analysis
  • Full removal of gas impurities for different applications
Gas purifiers are an essential part of your GC analysis as contaminants in gases can significantly impact the quality of results. Oxygen, hydrocarbons and moisture can lead to problems such as noisy baselines, moisture entering the GC column, excessive bleed and septa degradation. Even if carrier gas is of the highest quality, contaminants can be picked up from every part of the gas line. Therefore, a gas purifier is needed to ensure that maximum productivity is achieved.
Bestillingsoplysninger: Connecting units and filters must be ordered separately.
Leveringsinformation: Kits include gas purifiers and mounting unit.