tri-Natriumcitrat dihydrat Ph. Eur., Macron Fine Chemicals™
Leverandør: Avantor Performance Materials
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer: Sodium Citrate Dihydrate, Sodium citrate tribasic dihydrate, Sodium Citrate, Dihydrate, Sodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate, Citric acid trisodium salt dihydrate
P149-33EA 491 DKK
P149-33 P149-38
tri-Natriumcitrat dihydrat Ph. Eur., Macron Fine Chemicals™
tri-Natriumcitrat dihydrat
Formel: Na₃C₆H₅O₇·2H₂O
Molekylvægt: 294.1 g/mol
Smeltepunkt: 150
MDL Number: MFCD00150031
CAS-nummer: 6132-04-3
Merck Index: 13,08675
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e32147a215e8895f4b6db5aab1b99da667b795ac&vid=b929cda9d6f2eed18a8fefe57aa02b561af7c97f&prid=web&clid=SAPDAM