Saltsyre solution_SC 10% Biotech Reagent, meetSpec_Ph.Eur., Macron Fine Chemicals™
Leverandør: Avantor Performance Materials
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer: Hydrochloric acid solution, Muriatic acid
P111-33EA 221 DKK
P111-33 P111-38 P111-39 P111-40
Saltsyre solution_SC 10% Biotech Reagent, meetSpec_Ph.Eur., Macron Fine Chemicals™
Saltsyre
Formel: HCl
Molekylvægt: 36.46 g/mol
MDL Number: MFCD00011324
CAS-nummer: 7647-01-0
UN nummer: 1789
ADR: 8 II