di-Natriumhydrogenphosphat dodecahydrat Ph. Eur., Macron Fine Chemicals™
Leverandør: Avantor Performance Materials
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer: sec-Sodium phosphate, Sodium hydrogen phosphate dodecahydrate, Disodium phosphate, Dibasic sodium phosphate dodecahydrate, Sodium phosphate dibasic dodecahydrate, di-Sodium hydrogen phosphate dodecahydrate, Sodium monohydrogen phosphate dodecahydrate, di-Sodium hydrogen orthophosphate dodecahydrate
P116-33EA 562 DKK
P116-33 P116-42
di-Natriumhydrogenphosphat dodecahydrat Ph. Eur., Macron Fine Chemicals™
di-Natriumhydrogenphosphat dodecahydrat
Formel: Na₂HPO₄·12H₂O
Molekylvægt: 358.14 g/mol
Smeltepunkt: 35
MDL Number: MFCD00149181
CAS-nummer: 10039-32-4
Merck Index: 13,08733