Membrane disc filters, GLA-5000 PVC
Leverandør: Cytiva (Formerly Pall Lab)
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
66466 66467 66468 66469
514-0466EA 1800 DKK
514-0466 514-0467 514-0468 514-0469
Membrane disc filters, GLA-5000 PVC
Filtre Membranfiltre
Inherently low ash membrane ideally suited for multiple NIOSH analytical methods.
  • Assures gravimetric stability with low moisture pick-up and low tare weight
  • Low ash. Provides interference-free silica determinations
  • 5 μm pore size meets NIOSH and OSHA requirements
  • 25 and 37 mm sizes are ideal for use in Pall air monitoring cassettes
Excellent membrane choice for sampling airborne metals, silica, and dust.