Membrane disc filters, Supor® PES
Leverandør: Cytiva (Formerly Pall Lab)
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
60109 60110 60112 60114 60116 60170 60172 60173 60174 60175 60177 60179 60206 60298 60300 60301 60305 60307 60309 60310 60311 60334 61854 66234 66550 66551 66552 66553 66554 66555 66556 65472 60043
514-4151EA 1200 DKK
514-4151 514-4152 514-4153 514-4154 516-7739 514-4155 514-4156 514-4157 516-7743 514-4158 514-4159 516-7746 514-4160 514-4161 514-4162 514-4163 514-4164 516-7757 514-4165 514-4166 514-4167 514-4169 514-4174 514-4201 516-7932 516-7933 516-7934 516-7935 516-7936 516-7937 516-7938 516-7826 514-0377
Membrane disc filters, Supor® PES
Filtre Membranfiltre
Hydrophilic PES Supor® membrane for reduced filtration time, optimised for biological, pharmaceutical and sterilising filtration requirements.
  • Low protein binding and extensive drug compatibility for critical applications
  • Superior flow rates and high throughputs
Bestil nu
Specifikationer
Model Supor® Supor® Supor® Supor®
Environmentally Preferable
Porestørrelse 0,80 µm 0,45 µm 0,20 µm 0,10 µm
Thickness 140 µm 140 µm 145 µm 132 µm
Flowhastighed for vand 165 ml/min/cm² (0,7 bar) 58 ml/min/cm² (0,7 bar) 26 ml/min/cm² (0,7 bar) 5 ml/min/cm² (0,7 bar)
Max. temperature 100 °C 100 °C 100 °C 100 °C