TE buffer, 1X, ready to use, pH 8,0, low EDTA, Molecular Biology Grade, Affynmetrix/USB
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
J75793.AE J75793.AP
J75793.AEEA 787 DKK
J75793.AE J75793.AP
TE buffer, 1X, ready to use, pH 8,0, low EDTA, Molecular Biology Grade, Affynmetrix/USB
Buffere Biologiske buffere
TE buffer with low EDTA. Use directly to re-suspend and/or dilute purified DNA or RNA.
  • 1X TE solution consisting of 10 mM Tris Ultrapure, pH 8,0 and 0,1 mM EDTA
  • Solution is 0,2 μm filtered and dispensed into vials and/or durable, square Nalgene bottles