VWR®, Industrial Wipes, Raspy
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
115-2399EA 836 DKK
115-2399 115-2398
VWR®, Industrial Wipes, Raspy
Papirservietter Engangsservietter og - håndklæder Papirservietter til alment brug
These spunlace apertured wipes, made from 51% wood pulp and 49% polyester, are slightly abrasive on one side and soft on the other.
  • Extremely strong
  • Slightly abrasive side for heavy-duty action
  • Resistant to acids, solvents and chlorine
  • Recyclable
  • Food contact certified
Typical applications include: Food and beverage, automotive, industry, printing, laboratories, healthcare and janitorial tasks.
Basis weight: 75 g/m²
Absorption: 760%