VWR®, Industrial Wipes, Clean Smooth, Turquoise
Leverandør: VWR
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VWR®
115-2397EA 616 DKK
115-2397 115-2396
VWR®, Industrial Wipes, Clean Smooth, Turquoise
Papirservietter Engangsservietter og - håndklæder Papirservietter til alment brug
These spunlace wipes, made from 55% wood pulp and 45% polyester, are extremely strong, even when wet.
  • Low fibre and particle release
  • Resistant to acids and solvents
  • Good absorption
  • Do not contain any binders, silicone-free
  • Recyclable
  • Food contact certified
Typical applications include: Pharma, aerospace, semiconductor, food and beverage, automotive, laboratories, printing and healthcare.
Basis weight: 73 g/m²
Absorption: 600%