VWR®, Industrial Wipes, Clean Smooth, White
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
115-2394EA 1360 DKK
115-2394 115-2395
VWR®, Industrial Wipes, Clean Smooth, White
Papirservietter Engangsservietter og - håndklæder Papirservietter til alment brug
These bleached, spunlace wipes, made from 55% wood pulp and 45% polyester, are extremely strong, even when wet.
  • Low fibre and particle release
  • Resistant to acids, solvents and chlorine
  • Good absorption
  • Do not contain any binders, silicone-free
  • Recyclable
  • Food contact certified
Typical applications include: Pharma, aerospace, semiconductor, food and beverage, automotive, laboratories, printing and healthcare.
Basis weight: 68 g/m²
Absorption: 400%