VWR®, Cap Sealing Mats for 96 Deep Well Plates
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
732-3333EA 2000 DKK
732-3333 732-3334
VWR®, Cap Sealing Mats for 96 Deep Well Plates
Forsegling Mikrotiterpladetætninger
Square well cap mats for sealing 96-well shallow or deep well plates.
  • Choice of EVA or PP
  • Free from DNase and RNase