VWR®, Deep Well Plates, 96 Round Well
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
732-3322EA 1990 DKK
732-3322 732-3324 732-3328 732-3716 732-3718 732-3719 732-3802 732-3803
VWR®, Deep Well Plates, 96 Round Well
Mikroplader Deepwell Mikroplader
Made of virgin PP to minimise extractables.
  • Raised rim to prevent cross contamination and enable a better heat-welded seal
  • Also available rimless to allow insertion of vials, or when adhesive seals are used
  • Free from DNase and RNase
  • Round bottom for optimal mixing and recovery
  • Small volumes with flat, U- or V-bottoms are also available
  • Can be stored at –80 °C
Suitable for use with automated sample handling systems.