Wipes for general applications, WYPALL® X80
Leverandør: KIMBERLY CLARK
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
8377 8388
115-1987EA 1620 DKK
115-1987 115-0604
Wipes for general applications, WYPALL® X80
Papirservietter Genanvendelige rengøringsklude Klude til alment brug
WYPALL® X80 Cloth is fast absorbing thanks to the unique non woven HYDROKNIT® material. These bulky, reusable cloth-like wipes have an excellent absorption capacity.
  • Extra strong for the toughest wiping and cleaning tasks
  • Fast absorption
  • Tear resistant
Colour: Blue, white and red.