Sikkerhedsbriller, 2890-serien
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2890 2890A 2890SA 2890S
111-0433EA 218 DKK
111-0433 111-0434 111-0493 111-1636
Sikkerhedsbriller, 2890-serien
Beskyttelsesbriller
With a modern slim design, the 2800 series goggles combine performance, comfort and style while providing the wearer with an excellent field of vision. Available with PC or acetate lenses with or without indirect ventilation.
  • Easy to adjust strap
  • Pivoting ratchet for convenient individual adjustment and fit
  • Sealed (111-0493) or indirect ventilation
Weight: 99 g.
EN 166, EN 170
Certificeringer: EN 166, EN 170