Safety spectacles, A800
Leverandør: HONEYWELL SAFETY
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1015367 1015368 1015369 1015370
111-0399EA 55.3 DKK
111-0399 111-0517 111-0402 111-0166
Safety spectacles, A800
Briller
Single lens safety glasses for general protection against impact.
  • Temples with soft and padded inserts with extra flex built in
  • Slim profile for smaller faces
  • Sporty styling provides a fashionable fit
Suitable for applications in utilities, manufacturing, biotech, laboratories, cleanrooms, ship building and automotive manufacturing.
Certificeringer: EN 166.