Wipes for general applications, Tork Premium, colour coded
Leverandør: Essity
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
194450 194550 194650 194750
115-0584EA 1250 DKK
115-0584 115-0585 115-0586 115-0587
Wipes for general applications, Tork Premium, colour coded
Papirservietter Genanvendelige rengøringsklude Klude til alment brug
Low Linting, colour coded cloth wipes for cleaning, made from a mixture of viscose and polyester.
  • Useful for a colour coded cleaning system
  • Tork Premium: Strongest quality for heaviest wiping tasks