Wipes for general applications, Tork Low-Lint
Leverandør: Essity
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
190491 190494 190492 190493
SCAA190491EA 2090 DKK
SCAA190491 115-2078 115-2077 SCAA190493
Wipes for general applications, Tork Low-Lint
Papirservietter Engangsservietter og - håndklæder Papirservietter til alment brug
These Low Linting, flexible and drapable cloths work well with solvents, detergents and disinfectants. Available as a roll for use with W1/2/3 dispensers, folded for use with W8 dispensers or in a handy bucket dispenser (W10). Bucket allows cleaning product of choice to be added.
  • Multipurpose wipes: Handles most cleaning tasks
  • Low linting
  • Work well with solvents and detergents
  • Food contact approved
Colour: Turquoise.
Colour: Turquoise