Safety spectacles, 2840 series
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2840 2841
111-0432EA 185 DKK
111-0432 111-0520
Safety spectacles, 2840 series
Briller
These single lens safety glasses are available in different lens options: clear, yellow for improved contrast in low light conditions, smoke for protection against sun glare, light gold mirror for indoor/outdoor work, infra red shade five for certain gas welding/cutting and brazing applications and red-orange for protection against intense blue light.
  • Optical class 1 PC lens for highest optical clarity and extended wear time
  • Adjustable lens angle for individual fit
  • Adjustable temple length (4 positions) for wearer comfort and optimal fit
  • Integrated brow guard for increased protection
  • Soft inner padding on the temples for increased stability
Weight: 26 g
Certificeringer: EN 166, EN 170/172