Cleanroom face masks, BioClean DB™, PE
Leverandør: Nitritex
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BDBS-G BDBS
113-0783EA 7730 DKK
113-0783 113-0782
Cleanroom face masks, BioClean DB™, PE
Masker Renrumsmasker
The sterile pouch style mask has knitted headbands and all seams are ultrasonically bonded. The mask has inner and outer facing layers constructed of spunbond non woven polyester with the middle filter layer made from meltblown polyester.
  • Spacer maintains a clear breathing space for the wearer, improving the overall comfort and ease of breathing
  • Plastic coated, soft malleable steel nose bridge is fully enclosed
  • Available with or without integral neck guard to close the gap in protection between the coverall and throat of the wearer
Particle filtration efficiency: 0,3 µm 98,2%. Bacteria filtration efficiency: 3,0 µm 97,9%.
Suitable for use in class 10. ISO 4, M2.5 environments.
Certificeringer: Manufactured in a facility holding ISO 9001:2015. Processed in an NEBB certified ISO class 4 cleanroom environment. Sampling and inspection in accordance with ISO 2859-3:1991.
Emballering: 1 mask per inner bag; 20 inner bags per outer bag; 10 outer bags per carton (200 pieces).