Clip-on magnifiers, laboCLIP
Leverandør: ESCHENBACH OPTIK
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
164620 164625 164630 164640
630-8545EA 1350 DKK
630-8545 630-8546 630-8547 630-8548
Clip-on magnifiers, laboCLIP
Forstørrelsesglas og lupper
These clip-on magnifiers are designed to provide hands-free magnification, and fit most eyeglass frames.
  • Lightweight
  • Lens is set 2 inches from eye, allowing for a more comfortable working distance
  • Lens can be flipped up out of the way when not in use
Bridge = 25 mm
Jaws open = 15 mm
Bestillingsoplysninger: Replacement lenses are available and need to be ordered separately.