Combi safety storage cabinets, K-PHOENIX 90
Leverandør: asecos
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
30708001 30709001
MSGA30708001EA 29300 DKK
MSGA30708001 MSGA30709001
Combi safety storage cabinets, K-PHOENIX 90
Møbler Opbevaringskabinetter
Fireproof safety cabinets, classified type 90 (fire resistant up to 90 minutes), safe and approved storage of aggressive and flammable hazardous substances in working areas.
  • Robust construction and longevity - triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
  • Two in one - 2 compartments (vertical separation) allow the storage of flammable liquids and aggressive hazardous materials in one cabinet
  • User-friendly and safe (folding door compartment): door open arrest system (standard), convenient and safe removal or storage of containers
  • No unauthorised use - doors lockable with profi le cylinder (integration in an existing locking system possible) / cylinder lock
  • Easy alignment - adjusting aids to compensate for uneven floor
  • Ventilation - integrated air ducts ready for connection (DN 75) to a technical exhaust system
Cabinet has compartment with three shelves, perforated insert and bottom collecting sump of plastic laminated sheet steel and compartment with three tray shelves and bottom collecting sump of melamine/PP for the storage of flammable liquids.
Certificeringer: Furnace tested (type test) in accordance with EN 14470-1. EN 14727 conformity (laboratory furniture).
Bestillingsoplysninger: Interior equipment packages need to be ordered separately (not included). Please check the website or contact Avantor for further information.
Leveringsinformation: Delivery prices available upon request.