Lysing kits for Fastprep®-24 5G, Lysing Matrix M, MP Biomedicals
Leverandør: MP Biomedicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
116923500 116923100 116923050
ICNA116923500EA 7760 DKK
ICNA116923500 ICNA116923100 ICNA116923050
Lysing kits for Fastprep®-24 5G, Lysing Matrix M, MP Biomedicals
Homogenisatorer Homogenisatorer, bordmodeller
Each impact-resistant 2 ml tube contains one 6,35 mm diameter zirconium oxide coated ceramic grinding sphere.
  • Use to break hard/brittle samples
  • Stored at room temperatue (15 to 30 °C)
  • DNase and RNase free
Specialty applications include RNA extraction from difficult samples where lysis by impaction only is preferable with minimal amounts of mechanical shearing. This grinding matrix is chemically inert and will not bind nucleic acids.
Leveringsinformation: Includes an additional bag of grinding spheres.