Polyvalent alkaline detergent, RBS 35 Concentrate
Leverandør: CHEMICAL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
35CONC/E123 35CONC/E148
CHEP35CONC/E123EA 770 DKK
CHEP35CONC/E123 CHEP35CONC/E148
Polyvalent alkaline detergent, RBS 35 Concentrate
Renseopløsninger Detergenter og Overflade Rengøringsmidler
Suitable for efficient manual and ultrasonic cleaning of glassware and laboratory equipement made of stainless steel, plastic and porcelain.
  • High emulsifying and degreasing power
  • Low use concentration
  • Compatible with use in tap water
RBS 35 Concentrate is recommended for manual cleaning, soaking and ultrasonic cleaning of glassware and laboratory equipment made of glass, plastic, stainless steel and porcelain. It efficiently removes organic residues, blood and biological residues, fats, oils, distillation residues etc. It rinses easily without leaving any interfering residues. RBS 35 Concentrate can be used in tap water without risk of precipitation of scale.
Bestillingsoplysninger: Available in box of 12 × 1 litre, 4 × 5 litres, 20 kg drum. Please contact Avantor for details.
Advarsel: Not recommended for use on white metals.