Multipurpose alkaline detergent, RBS 25 Concentrate
Leverandør: CHEMICAL
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
25CONC/E032 25CONC/E148 25CONC/E123
CHEP25CONC/E032EA 195 DKK
CHEP25CONC/E032 CHEP25CONC/E148 CHEP25CONC/E123
Multipurpose alkaline detergent, RBS 25 Concentrate
Renseopløsninger Detergenter og Overflade Rengøringsmidler
Suitable for efficient manual and ultrasonic cleaning of glassware and laboratory equipement made of stainless steel, plastic and porcelain.
  • High wetting and emulsifying properties
  • Low use concentration
RBS 25 Concentrate is recommended for manual cleaning, soaking and ultrasonic cleaning of glassware and laboratory equipment made of glass, plastic, stainless steel and porcelain. It efficiently removes organic residues, blood and biological residues, fats, oils, distillation residues etc. It rinses easily without leaving any interfering residues.
Bestillingsoplysninger: Available in box of 12 × 1 litre, 4 x 5 litres, 20 kg drum. Please contact Avantor for details.
Advarsel: Not recommended for use on white metals.